Assay Details
Gene Symbol: bip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143666.3 | NP_651923.2 | 1 - 2 | 326 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
