Assay Details
Gene Symbol: bip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143666.3 | NP_651923.2 | 3 - 4 | 2530 | 74 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
