Assay Details
Gene Symbol: Thd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272131.2 | NP_001259060.1 | 4 - 5 | 2957 | 105 | |
NM_001297786.2 | NP_001284715.1 | 4 - 5 | 2957 | 105 | ||
NM_143668.3 | NP_651925.1 | 4 - 5 | 2957 | 105 | ||
GenBank mRNA | AJ277789.1 | - | 4 - 5 | 2911 | 105 | |
BT128699.1 | - | 3 - 4 | 2380 | 105 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
