Assay Details
Gene Symbol: Vha36-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143730.3 | NP_651987.1 | 1 | 925 | 150 | |
GenBank mRNA | AY069116.1 | - | 1 | 904 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
