Assay Details
| Gene Symbol: deltaCOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169171.2 | NP_001162642.1 | 3 - 4 | 461 | 52 | |
| NM_143755.4 | NP_652012.1 | 3 - 4 | 461 | 52 | ||
| GenBank mRNA | AY051528.1 | - | 3 - 4 | 463 | 52 | |
More Information
Set Membership: |
|

deltaCOP
Genomic Map