Assay Details
| Gene Symbol: Sec61beta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143780.4 | NP_652037.1 | 2 - 3 | 347 | 63 | |
| GenBank mRNA | AJ249754.1 | - | 2 - 3 | 320 | 63 | |
| BT050441.1 | - | 2 - 3 | 389 | 63 | ||
More Information
Set Membership: |
|

Sec61beta
Genomic Map