Assay Details
Gene Symbol: Reph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143795.4 | NP_652052.1 | 2 - 3 | 275 | 80 | |
NM_164561.3 | NP_722945.2 | 2 - 3 | 1229 | 80 | ||
GenBank mRNA | AY119477.1 | - | 2 - 3 | 1213 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_143795.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
