Assay Details
Gene Symbol: Reph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143795.4 | NP_652052.1 | 3 - 4 | 602 | 106 | |
NM_164561.3 | NP_722945.2 | 3 - 4 | 1556 | 106 | ||
GenBank mRNA | AY119477.1 | - | 3 - 4 | 1540 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
