Assay Details
| Gene Symbol: Hipk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382035.1 | NP_001369036.1 | 2 - 3 | 808 | 70 | |
| NM_138194.3 | NP_612038.2 | 2 - 3 | 730 | 70 | ||
| NM_167834.2 | NP_728531.1 | 2 - 3 | 808 | 70 | ||
More Information
Set Membership: |
|

Hipk
Genomic Map