Assay Details
Gene Symbol: Aplip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167856.3 | NP_728573.1 | 2 - 3 | 714 | 67 | |
GenBank mRNA | AF220194.1 | - | 2 - 3 | 692 | 67 | |
AF231037.1 | - | 2 - 3 | 625 | 67 | ||
AY051495.1 | - | 2 - 3 | 688 | 67 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
