Assay Details
Gene Symbol: Aplip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167856.3 | NP_728573.1 | 3 - 4 | 1438 | 76 | |
NM_167857.4 | NP_728574.1 | 3 - 4 | 1417 | 76 | ||
GenBank mRNA | AF220194.1 | - | 3 - 4 | 1416 | 76 | |
AF231037.1 | - | 3 - 4 | 1349 | 76 | ||
AY051495.1 | - | 3 - 4 | 1412 | 76 | ||
BT001795.1 | - | 3 - 4 | 1364 | 76 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
