Assay Details
Gene Symbol: Myo61F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057586.4 | NP_476934.2 | 6 - 7 | 3155 | 157 | |
NM_167870.2 | NP_728594.2 | 6 - 7 | 3232 | 157 | ||
NM_167871.2 | NP_728595.1 | 6 - 7 | 3240 | 157 | ||
NM_167872.2 | NP_728596.1 | 6 - 7 | 3378 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
