Assay Details
Gene Symbol: hfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169844.2 | NP_001163315.1 | 3 - 4 | 3478 | 68 | |
NM_001169845.2 | NP_001163316.1 | 5 - 6 | 1064 | 68 | ||
NM_001169846.2 | NP_001163317.1 | 6 - 7 | 1398 | 68 | ||
NM_001169847.2 | NP_001163318.1 | 4 - 5 | 3077 | 68 | ||
NM_001169848.2 | NP_001163319.1 | 5 - 6 | 1703 | 68 | ||
NM_001169849.2 | NP_001163320.1 | 5 - 6 | 1069 | 68 | ||
NM_080384.4 | NP_525123.2 | 4 - 5 | 1017 | 68 | ||
NM_167880.3 | NP_728610.1 | 5 - 6 | 1125 | 68 | ||
NM_167881.3 | NP_728611.1 | 5 - 6 | 1078 | 68 | ||
NM_167882.3 | NP_728612.1 | 7 - 8 | 1325 | 68 | ||
GenBank mRNA | AY061006.1 | - | 7 - 8 | 1324 | 68 | |
BT003317.1 | - | 3 - 4 | 3494 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
