Assay Details
| Gene Symbol: Gk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299975.1 | NP_001286904.1 | 2 - 3 | 970 | 61 | |
| NM_139398.3 | NP_647655.1 | 2 - 3 | 805 | 61 | ||
| NM_167912.2 | NP_728664.1 | 2 - 3 | 738 | 61 | ||
| NM_167913.3 | NP_728665.1 | 2 - 3 | 823 | 61 | ||
| NM_167914.2 | NP_728666.1 | 2 - 3 | 747 | 61 | ||
| NM_167915.3 | NP_728667.1 | 2 - 3 | 819 | 61 | ||
| GenBank mRNA | BT044165.1 | - | 2 - 3 | 788 | 61 | |
More Information
Set Membership: |
|

Gk2
Genomic Map