Assay Details
| Gene Symbol: Jafrac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274421.1 | NP_001261350.1 | 3 - 4 | 327 | 68 | |
| NM_080263.4 | NP_525002.1 | 3 - 4 | 367 | 68 | ||
| NM_167971.2 | NP_728793.1 | 3 - 4 | 498 | 68 | ||
| GenBank mRNA | AF167099.1 | - | 3 - 4 | 328 | 68 | |
| AF321614.1 | - | 3 - 4 | 343 | 68 | ||
| AY060785.1 | - | 3 - 4 | 343 | 68 | ||
More Information
Set Membership: |
|

Jafrac2
Genomic Map