Assay Details
| Gene Symbol: gry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139528.3 | NP_647785.3 | 7 - 8 | 2713 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gry| Gene Symbol: gry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139528.3 | NP_647785.3 | 7 - 8 | 2713 | 54 | |
Set Membership: |
|