Assay Details
Gene Symbol: Drsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168021.3 | NP_728862.1 | 1 | 185 | 112 | |
GenBank mRNA | AY338360.1 | - | 1 | 168 | 112 | |
AY351399.1 | - | 1 | 168 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
