Assay Details
| Gene Symbol: Drsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168027.1 | NP_728872.1 | 1 | 68 | 81 | |
| GenBank mRNA | AY351401.1 | - | 1 | 68 | 81 | |
| BT100068.4 | - | 1 | 68 | 81 | ||
| KX531554.1 | - | 1 | 68 | 81 | ||
More Information
Set Membership: |
|

Drsl1
Genomic Map