Assay Details
| Gene Symbol: RhoGEF64C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274496.1 | NP_001261425.1 | 9 - 10 | 6714 | 74 | |
| NM_168102.3 | NP_729032.1 | 10 - 11 | 6845 | 74 | ||
More Information
Set Membership: |
|

RhoGEF64C
Genomic Map