Assay Details
Gene Symbol: RhoGEF64C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274496.1 | NP_001261425.1 | 6 - 7 | 3061 | 77 | |
NM_168102.3 | NP_729032.1 | 7 - 8 | 3192 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
