Assay Details
Gene Symbol: Dhc64C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274498.1 | NP_001261427.1 | 10 - 11 | 9994 | 65 | |
NM_001274499.1 | NP_001261428.1 | 10 - 11 | 9982 | 65 | ||
NM_001274502.1 | NP_001261431.1 | 11 - 12 | 10021 | 65 | ||
NM_168103.3 | NP_729034.2 | 9 - 10 | 9955 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
