Assay Details
| Gene Symbol: form3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014571.1 | NP_001014571.1 | 8 - 9 | 3098 | 93 | |
| NM_001104057.1 | NP_001097527.1 | 9 - 10 | 3317 | 93 | ||
| GenBank mRNA | AB061681.1 | - | 8 - 9 | 3097 | 93 | |
More Information
Set Membership: |
|

form3
Genomic Map