Assay Details
| Gene Symbol: form3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014571.1 | NP_001014571.1 | 10 - 11 | 3800 | 85 | |
| NM_001104057.1 | NP_001097527.1 | 11 - 12 | 4019 | 85 | ||
| GenBank mRNA | AB061681.1 | - | 10 - 11 | 3799 | 85 | |
More Information
Set Membership: |
|

form3
Genomic Map