Assay Details
| Gene Symbol: Dscam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043123.3 | NP_001036588.2 | 6 - 7 | 1876 | 62 | |
| NM_001144424.2 | NP_001137896.1 | 6 - 7 | 1876 | 62 | ||
| NM_001144425.2 | NP_001137897.1 | 6 - 7 | 1876 | 62 | ||
| NM_001144426.2 | NP_001137898.1 | 6 - 7 | 1876 | 62 | ||
| NM_001169896.3 | NP_001163367.1 | 6 - 7 | 1876 | 62 | ||
| NM_001169897.2 | NP_001163368.1 | 6 - 7 | 1876 | 62 | ||
| NM_001274569.2 | NP_001261498.1 | 6 - 7 | 1876 | 62 | ||
| NM_001274570.2 | NP_001261499.1 | 6 - 7 | 1876 | 62 | ||
| NM_001274571.2 | NP_001261500.1 | 6 - 7 | 1876 | 62 | ||
| NM_001300035.1 | NP_001286964.1 | 6 - 7 | 1876 | 62 | ||
| NM_001300036.1 | NP_001286965.1 | 6 - 7 | 1876 | 62 | ||
| NM_001316478.1 | NP_001303407.1 | 6 - 7 | 1876 | 62 | ||
| NM_168194.4 | NP_729223.2 | 6 - 7 | 1876 | 62 | ||
| NM_168195.3 | NP_729224.2 | 6 - 7 | 1876 | 62 | ||
| NM_168196.4 | NP_729225.2 | 6 - 7 | 1876 | 62 | ||
| GenBank mRNA | BT133263.1 | - | 6 - 7 | 1863 | 62 | |
More Information
Set Membership: |
|

Dscam2
Genomic Map