Assay Details
Gene Symbol: Rpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132080.4 | NP_572308.3 | 3 - 4 | 1067 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
