Assay Details
Gene Symbol: Pat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272356.2 | NP_001259285.1 | 3 - 4 | 1136 | 75 | |
NM_001272357.2 | NP_001259286.1 | 3 - 4 | 1136 | 75 | ||
NM_132100.5 | NP_572328.1 | 3 - 4 | 1136 | 75 | ||
GenBank mRNA | AY058334.1 | - | 3 - 4 | 962 | 75 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
