Assay Details
Gene Symbol: shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298034.1 | NP_001284963.1 | 5 - 6 | 1309 | 68 | |
NM_001298035.1 | NP_001284964.1 | 5 - 6 | 1246 | 68 | ||
NM_001298036.1 | NP_001284965.1 | 5 - 6 | 1258 | 68 | ||
NM_132121.3 | NP_572349.1 | 5 - 6 | 1297 | 68 | ||
GenBank mRNA | AY051622.1 | - | 5 - 6 | 1297 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
