Assay Details
| Gene Symbol: C3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144694.2 | NP_001138166.1 | 13 - 14 | 4201 | 65 | |
| NM_001144695.3 | NP_001138167.1 | 11 - 12 | 4455 | 65 | ||
| NM_001144696.3 | NP_001138168.1 | 12 - 13 | 4608 | 65 | ||
| NM_001144697.2 | NP_001138169.1 | 12 - 13 | 4048 | 65 | ||
| NM_001272362.1 | NP_001259291.1 | 12 - 13 | 4452 | 65 | ||
| NM_001272363.1 | NP_001259292.1 | 11 - 12 | 3831 | 65 | ||
| NM_001272364.1 | NP_001259293.1 | 12 - 13 | 4608 | 65 | ||
| NM_001298037.1 | NP_001284966.1 | 11 - 12 | 4281 | 65 | ||
| NM_132122.5 | NP_572350.2 | 12 - 13 | 5271 | 65 | ||
| NM_176694.3 | NP_788867.1 | 12 - 13 | 5731 | 65 | ||
More Information
Set Membership: |
|

C3G
Genomic Map