Assay Details
| Gene Symbol: C3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144694.2 | NP_001138166.1 | 8 - 9 | 2554 | 58 | |
| NM_001144695.3 | NP_001138167.1 | 6 - 7 | 2808 | 58 | ||
| NM_001144696.3 | NP_001138168.1 | 7 - 8 | 2961 | 58 | ||
| NM_001144697.2 | NP_001138169.1 | 7 - 8 | 2401 | 58 | ||
| NM_001272362.1 | NP_001259291.1 | 7 - 8 | 2805 | 58 | ||
| NM_001272363.1 | NP_001259292.1 | 6 - 7 | 2184 | 58 | ||
| NM_001298037.1 | NP_001284966.1 | 6 - 7 | 2634 | 58 | ||
| NM_132122.5 | NP_572350.2 | 7 - 8 | 3624 | 58 | ||
| NM_176694.3 | NP_788867.1 | 7 - 8 | 4084 | 58 | ||
More Information
Set Membership: |
|

C3G
Genomic Map