Assay Details
Gene Symbol: Inx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132146.2 | NP_572374.1 | 1 - 2 | 954 | 63 | |
GenBank mRNA | AY057373.1 | - | 1 - 2 | 719 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
