Assay Details
| Gene Symbol: Atg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132162.4 | NP_572390.1 | 5 - 6 | 790 | 107 | |
| GenBank mRNA | AY069596.1 | - | 5 - 6 | 760 | 107 | |
More Information
Set Membership: |
|

Atg5
Genomic Map