Assay Details
| Gene Symbol: Gbeta5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132224.3 | NP_572452.1 | 1 | 1421 | 64 | |
| GenBank mRNA | BT004841.1 | - | 1 | 1426 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbeta5| Gene Symbol: Gbeta5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132224.3 | NP_572452.1 | 1 | 1421 | 64 | |
| GenBank mRNA | BT004841.1 | - | 1 | 1426 | 64 | |
Set Membership: |
|