Assay Details
Gene Symbol: PIP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132259.3 | NP_572487.3 | 1 - 2 | 388 | 102 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
