Assay Details
Gene Symbol: APC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169243.3 | NP_001162714.2 | 1 - 2 | 227 | 90 | |
NM_132294.3 | NP_572522.2 | 1 - 2 | 233 | 90 | ||
GenBank mRNA | BT133315.1 | - | 1 - 2 | 233 | 90 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
