Assay Details
| Gene Symbol: APC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169243.3 | NP_001162714.2 | 5 - 6 | 1205 | 64 | |
| NM_132294.3 | NP_572522.2 | 5 - 6 | 1217 | 64 | ||
| GenBank mRNA | BT133315.1 | - | 5 - 6 | 1217 | 64 | |
More Information
Set Membership: |
|

APC4
Genomic Map