Assay Details
| Gene Symbol: spz6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138117.3 | NP_611961.1 | 3 - 4 | 763 | 69 | |
| GenBank mRNA | BT001656.1 | - | 3 - 4 | 761 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spz6| Gene Symbol: spz6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138117.3 | NP_611961.1 | 3 - 4 | 763 | 69 | |
| GenBank mRNA | BT001656.1 | - | 3 - 4 | 761 | 69 | |
Set Membership: |
|