Assay Details
| Gene Symbol: CG44247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138130.4 | NP_611974.2 | 1 - 2 | 607 | 71 | |
| NM_166690.3 | NP_726489.2 | 1 - 2 | 607 | 71 | ||
| GenBank mRNA | BT100206.1 | - | 1 - 2 | 564 | 71 | |
More Information
Set Membership: |
|

CG44247
Genomic Map