Assay Details
Gene Symbol: NaCP60E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202078.3 | NP_001189007.2 | 26 - 27 | 8489 | 172 | |
NM_001259578.1 | NP_001246507.1 | 23 - 24 | 7590 | 172 | ||
NM_001259580.2 | NP_001246509.1 | 27 - 28 | 8504 | 172 | ||
NM_001274243.2 | NP_001261172.1 | 26 - 27 | 8489 | 172 | ||
NM_001274244.2 | NP_001261173.1 | 25 - 26 | 9694 | 172 | ||
NM_001274245.2 | NP_001261174.1 | 22 - 23 | 9596 | 172 | ||
NM_001274246.2 | NP_001261175.1 | 25 - 26 | 8596 | 172 | ||
NM_166696.5 | NP_726495.3 | 26 - 27 | 8752 | 172 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
