Assay Details
| Gene Symbol: Nplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043105.3 | NP_001036570.1 | 1 - 2 | 305 | 76 | |
| NM_001144288.2 | NP_001137760.1 | 1 - 2 | 305 | 76 | ||
| NM_138149.3 | NP_611993.1 | 1 - 2 | 305 | 76 | ||
| GenBank mRNA | AY060340.1 | - | 1 - 2 | 287 | 76 | |
| BT030135.1 | - | 1 - 2 | 288 | 76 | ||
More Information
Set Membership: |
|

Nplp1
Genomic Map