Assay Details
| Gene Symbol: Nplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043105.3 | NP_001036570.1 | 2 - 3 | 424 | 80 | |
| NM_001144288.2 | NP_001137760.1 | 2 - 3 | 424 | 80 | ||
| NM_138149.3 | NP_611993.1 | 2 - 3 | 424 | 80 | ||
| GenBank mRNA | AY060340.1 | - | 2 - 3 | 406 | 80 | |
| BT030135.1 | - | 2 - 3 | 407 | 80 | ||
More Information
Set Membership: |
|

Nplp1
Genomic Map