Assay Details
| Gene Symbol: Nplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138149.3 | NP_611993.1 | 3 - 4 | 1112 | 55 | |
| GenBank mRNA | AY060340.1 | - | 3 - 4 | 1094 | 55 | |
More Information
Set Membership: |
|

Nplp1
Genomic Map