Assay Details
Gene Symbol: MED30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138202.4 | NP_612046.1 | 1 | 1028 | 102 | |
GenBank mRNA | AY061627.1 | - | 1 | 1009 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
