Assay Details
| Gene Symbol: mRpL46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139404.3 | NP_647661.1 | 1 | 761 | 81 | |
| GenBank mRNA | AY071006.1 | - | 1 | 759 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mRpL46| Gene Symbol: mRpL46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139404.3 | NP_647661.1 | 1 | 761 | 81 | |
| GenBank mRNA | AY071006.1 | - | 1 | 759 | 81 | |
Set Membership: |
|