Assay Details
| Gene Symbol: Cpr62Bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144397.2 | NP_001137869.1 | 2 - 3 | 393 | 79 | |
| NM_001274363.1 | NP_001261292.1 | 3 - 4 | 327 | 79 | ||
| NM_139410.2 | NP_647667.1 | 3 - 4 | 305 | 79 | ||
| GenBank mRNA | BT010217.1 | - | 3 - 4 | 303 | 79 | |
More Information
Set Membership: |
|

Cpr62Bb
Genomic Map