Assay Details
| Gene Symbol: Mfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139422.3 | NP_647679.1 | 1 - 2 | 189 | 59 | |
| GenBank mRNA | AY047562.1 | - | 1 - 2 | 195 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfap1| Gene Symbol: Mfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139422.3 | NP_647679.1 | 1 - 2 | 189 | 59 | |
| GenBank mRNA | AY047562.1 | - | 1 - 2 | 195 | 59 | |
Set Membership: |
|