Assay Details
| Gene Symbol: ecd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139450.3 | NP_647707.1 | 1 - 2 | 367 | 75 | |
| GenBank mRNA | AY051483.1 | - | 1 - 2 | 293 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ecd| Gene Symbol: ecd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139450.3 | NP_647707.1 | 1 - 2 | 367 | 75 | |
| GenBank mRNA | AY051483.1 | - | 1 - 2 | 293 | 75 | |
Set Membership: |
|