Assay Details
Gene Symbol: ProRS-m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274408.1 | NP_001261337.1 | 1 - 2 | 201 | 66 | |
NM_139480.3 | NP_647737.1 | 1 - 2 | 201 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
