Assay Details
Gene Symbol: Pdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274615.1 | NP_001261544.1 | 5 - 6 | 1877 | 85 | |
NM_001274617.1 | NP_001261546.1 | 4 - 5 | 2778 | 85 | ||
NM_001274618.1 | NP_001261547.1 | 4 - 5 | 550 | 85 | ||
NM_001300044.1 | NP_001286973.1 | 4 - 5 | 1656 | 85 | ||
NM_080093.5 | NP_524832.3 | 4 - 5 | 1656 | 85 | ||
NM_168233.3 | NP_729297.1 | 3 - 4 | 508 | 85 | ||
NM_168235.3 | NP_729299.1 | 3 - 4 | 2745 | 85 | ||
NM_168236.3 | NP_729300.1 | 4 - 5 | 2787 | 85 | ||
NM_168237.2 | NP_729301.1 | 5 - 6 | 2382 | 85 | ||
NM_168238.3 | NP_729302.2 | 4 - 5 | 2323 | 85 | ||
NM_168239.2 | NP_729303.1 | 4 - 5 | 2426 | 85 | ||
GenBank mRNA | AF172402.2 | - | 3 - 4 | 891 | 85 | |
AF172403.2 | - | 4 - 5 | 933 | 85 | ||
AF172404.1 | - | 3 - 4 | 512 | 85 | ||
AY070526.1 | - | 5 - 6 | 2382 | 85 | ||
AY775786.1 | - | 4 - 5 | 612 | 85 | ||
BT003290.1 | - | 3 - 4 | 569 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
