Assay Details
Gene Symbol: ImpE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274648.1 | NP_001261577.1 | 2 - 3 | 508 | 69 | |
NM_080110.3 | NP_524849.2 | 2 - 3 | 452 | 69 | ||
NM_168270.2 | NP_729371.1 | 2 - 3 | 452 | 69 | ||
GenBank mRNA | AY095083.1 | - | 2 - 3 | 448 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
