Assay Details
Gene Symbol: Zasp66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169911.1 | NP_001163382.1 | 4 - 5 | 958 | 74 | |
NM_001169913.1 | NP_001163384.1 | 4 - 5 | 958 | 74 | ||
NM_001169914.3 | NP_001163385.1 | 4 - 5 | 969 | 74 | ||
NM_001169915.2 | NP_001163386.1 | 4 - 5 | 969 | 74 | ||
NM_139989.4 | NP_648246.3 | 4 - 5 | 958 | 74 | ||
NM_168283.4 | NP_729395.2 | 4 - 5 | 969 | 74 | ||
NM_168284.3 | NP_729396.2 | 4 - 5 | 958 | 74 | ||
NM_168286.4 | NP_729398.1 | 3 - 4 | 456 | 74 | ||
NM_176303.5 | NP_788481.1 | 3 - 4 | 456 | 74 | ||
NM_176304.5 | NP_788482.1 | 3 - 4 | 456 | 74 | ||
NM_176305.4 | NP_788483.1 | 3 - 4 | 456 | 74 | ||
NM_176306.3 | NP_788484.2 | 2 - 3 | 596 | 74 | ||
GenBank mRNA | AY058415.1 | - | 4 - 5 | 936 | 74 | |
BT029127.1 | - | 4 - 5 | 938 | 74 | ||
BT088849.1 | - | 3 - 4 | 432 | 74 | ||
BT089012.1 | - | 3 - 4 | 451 | 74 | ||
BT133470.1 | - | 4 - 5 | 967 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
